missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylformamide, Purified (DMF), Reagents
N,N-Dimethylformamide, Purified (DMF), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C2196200200E
Specifications
| N,N-Dimethylformamide, Purified (DMF) | |
| 68-12-2 | |
| 152°C to 153°C | |
| Steel Drum | |
| 0.944 | |
| Dimethylformamide, DMF, N,N-Dimethylmethanamide | |
| CN(C)C=O | |
| 73.10 |
| -60.5°C | |
| Colorless to Slight Yellow | |
| 200 L | |
| Liquid | |
| C3H7NO | |
| ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
| N,N-dimethylformamide | |
| Pure |
Chemical Identifiers
| 68-12-2 | |
| 73.10 | |
| Dimethylformamide, DMF, N,N-Dimethylmethanamide | |
| CN(C)C=O |
| C3H7NO | |
| ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
| N,N-dimethylformamide |