missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, Purified (DMF), Reagents

Catalog No. p-200021084
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337283 1 L
01337282 200 L
01337281 20 L
01337280 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337283 Supplier Labchem Inc Supplier No. C21962001C
Only null left

N,N-Dimethylformamide, Purified (DMF), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym Dimethylformamide, DMF, N,N-Dimethylmethanamide
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Melting Point -60.5°C
Color Colorless to Slight Yellow
Boiling Point 152°C to 153°C
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.944
Grade Pure
Chemical Name or Material N,N-Dimethylformamide, Purified (DMF)