missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, For pesticide residue analysis, Honeywell Riedel-de Haen

CHROMASOLV™ for pesticide residue analysis

$770.00 - $770.00

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6001002
View Documents
Honeywell Chemicals
3448925L
2.5 L
Each for $770.00
Only null left
 
Description

Description

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
Specifications

Chemical Identifiers

68-12-2
73.10
ZMXDDKWLCZADIW-UHFFFAOYSA-N
6228
N,N-dimethylformamide
C3H7NO
MFCD00003284
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
CHEBI:17741
CN(C)C=O

Specifications

68-12-2
2.5 L
58°C (136.4°F)
C3H7NO
MFCD00003284
UN2265
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
CN(C)C=O
73.10
CHEBI:17741
≥99.8% (GC)
N,N-Dimethylformamide
0.944g/mL
0.152
833°F
HCON(CH3)2
2.7 mmHg (20°C)
605365
ZMXDDKWLCZADIW-UHFFFAOYSA-N
N,N-dimethylformamide
6228
73.09g/mol
CHROMASOLV
Videos
Safety and Handling

Safety and Handling

P201-P280-P305 + P351 + P338-P308 + P313

RTECSNumber : LQ2100000

SDS
missing translation for 'documents'

missing translation for 'documents'