missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, For pesticide residue analysis, Solstice

Catalog No. 6001002
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001002 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6001002 Supplier Solstice Supplier No. 344892.5L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CHROMASOLV™ for pesticide residue analysis

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Density 0.944g/mL
Quantity 2.5 L
Explosion Limit 0.152
Flash Point 58°C (136.4°F)
Ignition Point 833°F
Linear Formula HCON(CH3)2
Vapor Pressure 2.7 mmHg (20°C)
UN Number UN2265
Beilstein 605365
Formula Weight 73.09g/mol
Percent Purity ≥99.8% (GC)
Grade CHROMASOLV
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less
Hazard Statement P201-P280-P305 + P351 + P338-P308 + P313
RTECSNumber LQ2100000