missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide Dineopentyl Acetal 96.0+%, TCI America™

Catalog No. p-7126814
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15955ML 5 mL
D159525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15955ML Supplier TCI America Supplier No. D15955ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4909-78-8
Molecular Formula C13H30NO2
Molecular Weight (g/mol) 232.39
MDL Number MFCD00008851
InChI Key KEXFRBIOHPDZQM-UHFFFAOYSA-O
Synonym n,n-dimethylformamide dineopentyl acetal, methanamine, 1,1-bis 2,2-dimethylpropoxy-n,n-dimethyl, n,n-dimethyl-1,1-bis neopentyloxy methanamine, dimethylformamide dineopentyl acetal, 1,1-bis 2,2-dimethylpropoxy-n,n-dimethylmethanamine, bis 2,2-dimethylpropoxy methyl dimethylamine, acmc-1al8s, 1,1-bis 2,2-dimethylpropoxy-n,n,n-trimethylamine, 1,1-dineopentyloxytrimethylamine, dimethylformamide dineopentylacetal
PubChem CID 78623
IUPAC Name [bis(2,2-dimethylpropoxy)methyl]dimethylazanium
SMILES C[NH+](C)C(OCC(C)(C)C)OCC(C)(C)C

Specifications

Color Colorless
Boiling Point 87°C
Quantity 5 mL
Formula Weight 231.38
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylformamide Dineopentyl Acetal [for Esterification]