accessibility menu, dialog, popup

UserName

CH3NHH3COCH3CH3H3COCH3CH3H3C

CAS RN 4909-78-8

IUPAC Name: [bis(2,2-dimethylpropoxy)methyl]dimethylazanium
Synonyms: [bis(2,2-dimethylpropoxy)methyl]dimethylazanium
Molecular Weight (g/mol): 232.39
Molecular Formula: C13H30NO2
InChi Key: KEXFRBIOHPDZQM-UHFFFAOYSA-O
SMILES: C[NH+](C)C(OCC(C)(C)C)OCC(C)(C)C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)