missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, ACS, 99.8+%

A solvent For Chromogenic Substrates | CAS: 68-12-2 | C3H7NO | 73.10 g/mol

$82.26 - $340.70

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA39117AP
View Documents
Thermo Scientific Chemicals
039117AP
500 mL
Each for $82.26
Only null left
 
AA39117K2
View Documents
Thermo Scientific Chemicals
039117K2
1 L
Each for $113.15
Only null left
 
AA39117M1
View Documents
Thermo Scientific Chemicals
039117M1
4 x 1 L
Each for $340.70
Only null left
 
Description

Description

N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Maximum level of impurities
  • Appearance Clear
  • Color (APHA) 15
  • Evaporation residue 0.005%
  • Titratable base 0.003meq/g
  • Titratable acid 0.0005meq/g
  • H2O 0.15%
Specifications

Chemical Identifiers

68-12-2
73.10
ZMXDDKWLCZADIW-UHFFFAOYSA-N
6228
N,N-dimethylformamide
C3H7NO
MFCD00003284
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
CHEBI:17741
CN(C)C=O

Specifications

68-12-2
Colorless
153°C
440°C (824°F)
Amine-like
1.431
500 mL
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
ZMXDDKWLCZADIW-UHFFFAOYSA-N
N,N-dimethylformamide
6228
73.09
ACS
N, N-Dimethylformamide, For analysis ACS
°C to 61°C
0.944 g/cm3 at 20°C
57°C (135°F)
>99.8%
C3H7NO
MFCD00003284
605365
Miscible with water and most organic solvents.
CN(C)C=O
73.10
CHEBI:17741
3.5 hPa (3mm Hg) at 20°C
Liquid
Videos
Safety and Handling

Safety and Handling

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P312-P363-P370+P378q-P501c

H226-H312+H332-H319-H335-H336-H350-H360D

DOTInformation : DOT Class: 3, Packaging Group: III

EINECSNumber : 200-679-5

RTECSNumber : LQ2100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only