Learn More
N,N-Dimethylformamide, ACS, 99.8+%
A solvent For Chromogenic Substrates | CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Supplier: Thermo Scientific Chemicals 039117K2
Description
N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Maximum level of impurities- Appearance Clear
- Color (APHA) 15
- Evaporation residue 0.005%
- Titratable base 0.003meq/g
- Titratable acid 0.0005meq/g
- H2O 0.15%
Specifications
| N, N-Dimethylformamide | |
| 68-12-2 | |
| Colorless | |
| 153°C | |
| 440°C (824°F) | |
| Amine-like | |
| 1.431 | |
| 1 L | |
| dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
| ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
| N,N-dimethylformamide | |
| 6228 | |
| 73.09 | |
| ACS |
| For analysis ACS | |
| °C to 61°C | |
| 0.944 g/cm3 at 20°C | |
| 57°C (135°F) | |
| >99.8% | |
| C3H7NO | |
| MFCD00003284 | |
| 605365 | |
| Miscible with water and most organic solvents. | |
| CN(C)C=O | |
| 73.10 | |
| CHEBI:17741 | |
| 3.5 hPa (3mm Hg) at 20°C | |
| Liquid |
Chemical Identifiers
| 68-12-2 | |
| 73.10 | |
| ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
| 6228 | |
| N,N-dimethylformamide |
| C3H7NO | |
| MFCD00003284 | |
| dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
| CHEBI:17741 | |
| CN(C)C=O |
Safety and Handling
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P312-P363-P370+P378q-P501c
H226-H312+H332-H319-H335-H336-H350-H360D
DOTInformation : DOT Class: 3, Packaging Group: III
EINECSNumber : 200-679-5
RTECSNumber : LQ2100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only