missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylethylamine 98.0+%, TCI America™

Catalog No. D1538500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1538500ML 500 mL
D153825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1538500ML Supplier TCI America Supplier No. D1538500ML

Chemical Identifiers

CAS 598-56-1
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.139
MDL Number MFCD00009039
InChI Key DAZXVJBJRMWXJP-UHFFFAOYSA-N
Synonym n,n-dimethylethylamine, dimethylethylamine, ethanamine, n,n-dimethyl, ethyldimethylamine, n-ethyldimethylamine, ethylamine, n,n-dimethyl, methanamine, n-ethyl-n-methyl, unii-9n5384xvem, c13-15-alkyldimethylamines, amines, c13-15-alkyldimethyl
PubChem CID 11723
IUPAC Name N,N-dimethylethanamine
SMILES CCN(C)C

Specifications

Color Colorless
Boiling Point 37°C
Quantity 500 mL
UN Number 2733
Formula Weight 73.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylethylamine
EINECSNumber (2)-3389
RTECSNumber KH2348550
TSCA Yes