accessibility menu, dialog, popup

UserName

H3CNCH3H3C

CAS RN 598-56-1

IUPAC Name: ethyldimethylamine
Synonyms: ethyl dimethylamine
Molecular Weight (g/mol): 73.14
Molecular Formula: C4H11N
InChi Key: DAZXVJBJRMWXJP-UHFFFAOYSA-N
SMILES: CCN(C)C
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)