missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylethylamine, 99%

Catalog No. p-7027324
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB23992AE 100 mL
AAB23992AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB23992AE Supplier Thermo Scientific Chemicals Supplier No. B23992AE

CAS: 598-56-1 | C4H11N | 73.139 g/mol

N,N-Dimethylethylamine is used as adhesive, sealant chemical and process regulator. It acts as an intermediate for quaternary ammonium compounds and is also used as a stabilizer for chlorinated hydrocarbons and vinyl derivatives. It is used in foundries as a catalyst for sand core production. Further, it is employed in the polymerization of polyamides. In addition to this, it is used in the preparation of polypeptide nanogels with self-reinforced endocytoses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylethylamine is used as adhesive, sealant chemical and process regulator. It acts as an intermediate for quaternary ammonium compounds and is also used as a stabilizer for chlorinated hydrocarbons and vinyl derivatives. It is used in foundries as a catalyst for sand core production. Further, it is employed in the polymerization of polyamides. In addition to this, it is used in the preparation of polypeptide nanogels with self-reinforced endocytoses.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents and peroxide.

Chemical Identifiers

CAS 598-56-1
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.139
MDL Number MFCD00009039
InChI Key DAZXVJBJRMWXJP-UHFFFAOYSA-N
Synonym n,n-dimethylethylamine, dimethylethylamine, ethanamine, n,n-dimethyl, ethyldimethylamine, n-ethyldimethylamine, ethylamine, n,n-dimethyl, methanamine, n-ethyl-n-methyl, unii-9n5384xvem, c13-15-alkyldimethylamines, amines, c13-15-alkyldimethyl
PubChem CID 11723
IUPAC Name N,N-dimethylethanamine
SMILES CCN(C)C

Specifications

Melting Point -140°C
Density 0.675
Boiling Point 36°C to 37°C
Flash Point −36°C (−33°F)
Odor Ammonia-like
Refractive Index 1.373
Linear Formula CH3CH2N(CH3)2
Quantity 100 mL
UN Number UN2733
Beilstein 1696893
Sensitivity Air sensitive
Solubility Information Miscible with water.
Formula Weight 73.14
Percent Purity 99%
Chemical Name or Material N,N-Dimethylethylamine
Show More Show Less
Hazard Category H225-H302+H332-H314
Hazard Statement GHS H Statement
H225-H314-H318-H302-H332
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 209-940-8
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only