Learn More
N,N-Dimethylethylamine, 99%
CAS: 598-56-1 | C4H11N | 73.13 g/mol
$203.38 - $203.38
Chemical Identifiers
| CAS | 598-56-1 |
|---|---|
| Molecular Formula | C4H11N |
| Molecular Weight (g/mol) | 73.13 |
| MDL Number | MFCD00009039 |
| InChI Key | DAZXVJBJRMWXJP-UHFFFAOYSA-N |
| Synonym | n,n-dimethylethylamine, dimethylethylamine, ethanamine, n,n-dimethyl, ethyldimethylamine, n-ethyldimethylamine, ethylamine, n,n-dimethyl, methanamine, n-ethyl-n-methyl, unii-9n5384xvem, c13-15-alkyldimethylamines, amines, c13-15-alkyldimethyl |
| PubChem CID | 11723 |
| IUPAC Name | N,N-dimethylethanamine |
| SMILES | CCN(C)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC219292500
|
Thermo Scientific Chemicals
219292500 |
250 mL | Glass bottle |
Each for $203.38
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 598-56-1 | |
| 73.13 | |
| DAZXVJBJRMWXJP-UHFFFAOYSA-N | |
| 11723 | |
| CCN(C)C |
| C4H11N | |
| MFCD00009039 | |
| n,n-dimethylethylamine, dimethylethylamine, ethanamine, n,n-dimethyl, ethyldimethylamine, n-ethyldimethylamine, ethylamine, n,n-dimethyl, methanamine, n-ethyl-n-methyl, unii-9n5384xvem, c13-15-alkyldimethylamines, amines, c13-15-alkyldimethyl | |
| N,N-dimethylethanamine |
Specifications
| 598-56-1 | |
| 100.0 | |
| Colorless to Yellow | |
| 36°C | |
| Authentic | |
| Glass bottle | |
| 1.3715 to 1.3735 | |
| MFCD00009039 | |
| 04, 94 | |
| n,n-dimethylethylamine, dimethylethylamine, ethanamine, n,n-dimethyl, ethyldimethylamine, n-ethyldimethylamine, ethylamine, n,n-dimethyl, methanamine, n-ethyl-n-methyl, unii-9n5384xvem, c13-15-alkyldimethylamines, amines, c13-15-alkyldimethyl | |
| DAZXVJBJRMWXJP-UHFFFAOYSA-N | |
| N,N-dimethylethanamine | |
| 11723 | |
| 99% | |
| N, N-Dimethylethylamine, 99% |
| 98.5 | |
| -140°C | |
| 0.6750g/mL | |
| -36°C | |
| 99% | |
| C4H11N | |
| C2H5N(CH3)2 | |
| 250 mL | |
| 0.675 | |
| Solubility in water: soluble. Other solubilities: miscible with most common organic solvents | |
| CCN(C)C | |
| 73.13 | |
| 73.13 | |
| Liquid |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapour.
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth
GHS Signal Word: Danger
EINECSNumber : 209-940-8
TSCA : TSCA
RUO – Research Use Only