missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylbutyramide, 98%

Catalog No. AAL0555606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0555606 5 g
AAL0555614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0555606 Supplier Thermo Scientific Chemicals Supplier No. L0555606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 760-79-2 | C6H13NO | 115.176 g/mol

N,N-Dimethylbutyramide, is used as an organic intermediate, pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylbutyramide, is used as an organic intermediate, pharmaceutical intermediate.

Solubility
Very Soluble in water (110 g/L) (25°C).

Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 760-79-2
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00015222
InChI Key VIJUZNJJLALGNJ-UHFFFAOYSA-N
Synonym n,n-dimethylbutyramide, butanamide, n,n-dimethyl, n,n-dimethyl-n-butyramide, nn-dimethylbutyramide, butyramide, n,n-dimethyl, butanamide,n-dimethyl, butanamide,n,n-dimethyl, butanoic acid dimethylamide, nn-dimethylbutyramide, pract., wln: 3vn1&1
PubChem CID 12973
IUPAC Name N,N-dimethylbutanamide
SMILES CCCC(=O)N(C)C

Specifications

Boiling Point 80°C to 82°C (15 mmHg)
Refractive Index 1.4425
Quantity 5 g
Beilstein 1742172
Solubility Information Very Soluble in water (110g/L) (25°C).
Formula Weight 115.17
Percent Purity 98%
Chemical Name or Material N,N-Dimethylbutyramide
RTECSNumber ES4319000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only