accessibility menu, dialog, popup

UserName

CAS RN 760-79-2

IUPAC Name: N,N-dimethylbutanamide
Synonyms: butanamide, N,N-dimethyl-
Molecular Weight (g/mol): 115.18
Molecular Formula: C6H13NO
InChi Key: VIJUZNJJLALGNJ-UHFFFAOYSA-N
SMILES: CCCC(=O)N(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results