missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylbenzylamine 98.0+%, TCI America™

Catalog No. p-7126375
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D068825ML 25 mL
D0688500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D068825ML Supplier TCI America Supplier No. D068825ML
Only null left

Chemical Identifiers

CAS 103-83-3
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00008329
InChI Key XXBDWLFCJWSEKW-UHFFFAOYSA-N
Synonym n,n-dimethylbenzylamine, benzyldimethylamine, n-benzyldimethylamine, dimethylbenzylamine, bdma, benzenemethanamine, n,n-dimethyl, benzyl-n,n-dimethylamine, n-phenylmethyl dimethylamine, n,n-dimethylbenzenemethanamine, araldite accelerator 062
PubChem CID 7681
IUPAC Name benzyldimethylamine
SMILES CN(C)CC1=CC=CC=C1

Specifications

Melting Point -75°C
Color Yellow
Boiling Point 180°C
Quantity 25 mL
UN Number 2619
Formula Weight 135.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylbenzylamine
EINECSNumber (3)-0332&(3)-0350
RTECSNumber DP4500000
TSCA Yes