missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylbenzamide 99.0+%, TCI America™

Catalog No. D025625G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D025625G 25 g
D0256100G 100 g
D0256500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D025625G Supplier TCI America Supplier No. D025625G

Chemical Identifiers

CAS 611-74-5
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00008320
InChI Key IMNDHOCGZLYMRO-UHFFFAOYSA-N
Synonym dimethylbenzamide, benzamide, n,n-dimethyl, nn-dimethylbenzamide, dimethylbenzmide, n,n-dmethylbenzamde, dmbz, n,n-dimethyl benzamide, n,n-dimethyl-benzamide, acmc-209mps, benzamide,n,n-dimethyl
PubChem CID 11916
IUPAC Name N,N-dimethylbenzamide
SMILES CN(C)C(=O)C1=CC=CC=C1

Specifications

Melting Point 43°C
Color White
Quantity 25 g
Formula Weight 149.19
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material N,N-Dimethylbenzamide
RTECSNumber CV4500000
TSCA Yes