accessibility menu, dialog, popup

UserName

CH3NH3CO

CAS RN 611-74-5

IUPAC Name: N,N-dimethylbenzamide
Synonyms: N,N-dimethylbenzamide
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: IMNDHOCGZLYMRO-UHFFFAOYSA-N
SMILES: CN(C)C(=O)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)