missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylbenzamide, 98+%

Catalog No. AAA1876636
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1876636 500 g
AAA1876614 25 g
AAA1876622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1876636 Supplier Thermo Scientific Chemicals Supplier No. A1876636
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 611-74-5 | C9H11NO | 149.19 g/mol

Reagent for deoxygenation of secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for deoxygenation of secondary alcohols.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 611-74-5
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00008320
InChI Key IMNDHOCGZLYMRO-UHFFFAOYSA-N
Synonym dimethylbenzamide, benzamide, n,n-dimethyl, nn-dimethylbenzamide, dimethylbenzmide, n,n-dmethylbenzamde, dmbz, n,n-dimethyl benzamide, n,n-dimethyl-benzamide, acmc-209mps, benzamide,n,n-dimethyl
PubChem CID 11916
IUPAC Name N,N-dimethylbenzamide
SMILES CN(C)C(=O)C1=CC=CC=C1

Specifications

Melting Point 42°C to 45°C
Boiling Point 132°C to 133°C (15 mmHg)
Flash Point >110°C (230°F)
Assay Percent Range 0.98
Quantity 500 g
Beilstein 774955
Solubility Information Soluble in water.
Formula Weight 149.19
Percent Purity ≥98%
Chemical Name or Material N,N-Dimethylbenzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-279-2
RTECSNumber CV4500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only