missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylaniline 99.0+%, TCI America™

Catalog No. D38661G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38661G 1 g
D38665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38661G Supplier TCI America Supplier No. D38661G

Chemical Identifiers

CAS 121-69-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00008304
InChI Key JLTDJTHDQAWBAV-UHFFFAOYSA-N
Synonym dimethylaniline, dimethylphenylamine, n,n-dimethylbenzenamine, benzenamine, n,n-dimethyl, dimethylamino benzene, n,n-dimethylphenylamine, n,n-dimethylbenzeneamine, dimethylaminobenzene, dwumetyloanilina, n,n-dimethylamino benzene
PubChem CID 949
ChEBI CHEBI:16269
IUPAC Name N,N-dimethylaniline
SMILES CN(C)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 194°C
Quantity 1 g
UN Number 2253
Formula Weight 121.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylaniline [for Biochemical Research]
EINECSNumber (3)-0114&(3)-0129
RTECSNumber BX4725000
TSCA Yes