missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylaniline, 99%

Catalog No. AAA11916AE
Change view
Click to view available options
Quantity:
10,000 mL
100 mL
500 mL
2500 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA11916AE 100 mL
AAA11916AP 500 mL
AAA119160F 2500 mL
AAA119160D 10,000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA11916AE Supplier Thermo Scientific Chemicals Supplier No. A11916AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 121-69-7 | C8H11N | 121.18 g/mol

N,N-Dimethylaniline acts as a precursor to triarylmethane dyes such as crystal violet and malachite green. It is also used in the synthesis of vanillin, methyl violet, and Michler's ketone. Further, it is used as a hardener for plastic resins and an acid scavenger in the manufacture of semisynthetic penicillins and cephalosporins. In addition to this, it is used as a promoter in the curing of polyester and vinyl ester resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylaniline acts as a precursor to triarylmethane dyes such as crystal violet and malachite green. It is also used in the synthesis of vanillin, methyl violet, and Michler′s ketone. Further, it is used as a hardener for plastic resins and an acid scavenger in the manufacture of semisynthetic penicillins and cephalosporins. In addition to this, it is used as a promoter in the curing of polyester and vinyl ester resins.

Solubility
Miscible with water, ethanol, ether, acetone, chloroform, benzene and alkaline.

Notes
Incompatible with strong oxidizing agents, Strong acids, acid chlorides, acid anhydrides, chloroformates and halogens.

Chemical Identifiers

CAS 121-69-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00008304
InChI Key JLTDJTHDQAWBAV-UHFFFAOYSA-N
Synonym dimethylaniline, dimethylphenylamine, n,n-dimethylbenzenamine, benzenamine, n,n-dimethyl, dimethylamino benzene, n,n-dimethylphenylamine, n,n-dimethylbenzeneamine, dimethylaminobenzene, dwumetyloanilina, n,n-dimethylamino benzene
PubChem CID 949
ChEBI CHEBI:16269
IUPAC Name N,N-dimethylaniline
SMILES CN(C)C1=CC=CC=C1

Specifications

Melting Point 2°C
Density 0.956
pH 7.4
Boiling Point 193°C
Flash Point 62°C (143°F)
Odor Characteristic
Assay Percent Range 99%
Refractive Index 1.558
Quantity 100 mL
UN Number UN2253
Beilstein 507140
Merck Index 14,3234
Solubility Information Miscible with water,ethanol,ether,acetone,chloroform,benzene and alkaline.
Formula Weight 121.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N,N-Dimethylaniline
Show More Show Less
Hazard Category H227-H301+H311+H331-H351
Hazard Statement GHS H Statement
H301-H311-H331-H351-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing cancer.
Combustible liquid.
Precautionary Statement P201-P202-P210-P235-P261-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: N,N-DIMETHYLANILINE
EINECSNumber 204-493-5
RTECSNumber BX4725000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only