missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylaniline, 99%, Spectrum™ Chemical

Catalog No. 18611860
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611860 100 mL Amber Glass Bottle
18611863 500 mL Amber Glass Bottle
18611861 2.5 L Amber Glass Bottle
18611862 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18611860 Supplier Spectrum Chemical Mfg Cor Supplier No. D1025100ML
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 121-69-7

Specifications

CAS Max % 1
Density 0.956 g/mL
Boiling Point 194°C / 381.2°F
Packaging Amber Glass Bottle
Quantity 100 mL
Molecular Weight (g/mol) 121.18
Percent Purity 0.99
Grade Reagent
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material N,N-Dimethylaniline

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.