missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylallylamine, 98%

Catalog No. p-115947
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC407980050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC407980050 Supplier Thermo Scientific Chemicals Supplier No. 407980050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2155-94-4 | C5H11N | 85.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2155-94-4
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD00038234
InChI Key GBCKRQRXNXQQPW-UHFFFAOYSA-N
Synonym n,n-dimethylallylamine, allyldimethylamine, dimethylallylamine, 2-propen-1-amine, n,n-dimethyl, 1-dimethylamino-2-propene, n-allyl-n,n-dimethylamine, n-allyldimethylamine, dimethyl prop-2-en-1-yl amine, allylamine, n,n-dimethyl, ch2=chch2n ch3 2
PubChem CID 75082
IUPAC Name N,N-dimethylprop-2-en-1-amine
SMILES CN(C)CC=C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.7410g/mL
Boiling Point 62°C
Flash Point 8°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4000 to 1.402
Linear Formula CH2CHCH2N(CH3)2
Quantity 5 g
Specific Gravity 0.741
Solubility Information Solubility in water: soluble
Formula Weight 85.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N, N-Dimethylallylamine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call
EINECSNumber 218-458-7
TSCA TSCA

RUO – Research Use Only