accessibility menu, dialog, popup

UserName

CAS RN 2155-94-4

IUPAC Name: dimethyl(prop-2-en-1-yl)amine
Synonyms: allyldimethyl amine
Molecular Weight (g/mol): 85.15
Molecular Formula: C5H11N
InChi Key: GBCKRQRXNXQQPW-UHFFFAOYSA-N
SMILES: CN(C)CC=C
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)