missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylacetamide Dimethyl Acetal (stabilized with 5-10% Methanol) 90.0+%, TCI America™
Supplier: TCI America D165225ML
Specifications
| N,N-Dimethylacetamide Dimethyl Acetal (stabilized with 5-10% Methanol) | |
| Yellow | |
| MFCD00008476 | |
| 1993 | |
| FBZVZUSVGKOWHG-UHFFFAOYSA-N | |
| 1,1-dimethoxy-N,N-dimethylethanamine | |
| 87835 | |
| ≥90.0% (T) |
| 18871-66-4 | |
| C6H15NO2 | |
| 25 mL | |
| n,n-dimethylacetamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylethylamine, 1,1-dimethoxyethyl dimethylamine, 1,1-dimethoxyethyl dimethyl amine, 1,1-dimethoxy-ethyl-dimethyl-amine, ethanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylacetamide dimethylacetal, dimethylacetamide dimethyl acetal, n,n-dimethylacetamide dimethyl acetal, stabilized with methanol, zlchem 332 | |
| CC(N(C)C)(OC)OC | |
| 133.191 | |
| 133.19 | |
| Liquid |
Chemical Identifiers
| 18871-66-4 | |
| 133.191 | |
| FBZVZUSVGKOWHG-UHFFFAOYSA-N | |
| 87835 | |
| CC(N(C)C)(OC)OC |
| C6H15NO2 | |
| MFCD00008476 | |
| n,n-dimethylacetamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylethylamine, 1,1-dimethoxyethyl dimethylamine, 1,1-dimethoxyethyl dimethyl amine, 1,1-dimethoxy-ethyl-dimethyl-amine, ethanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylacetamide dimethylacetal, dimethylacetamide dimethyl acetal, n,n-dimethylacetamide dimethyl acetal, stabilized with methanol, zlchem 332 | |
| 1,1-dimethoxy-N,N-dimethylethanamine |
Safety and Handling
TSCA : No