accessibility menu, dialog, popup

UserName

CAS RN 18871-66-4

IUPAC Name: (1,1-dimethoxyethyl)dimethylazanium
Synonyms: (1,1-dimethoxyethyl)dimethylazanium
Molecular Weight (g/mol): 134.2
Molecular Formula: C6H16NO2
InChi Key: FBZVZUSVGKOWHG-UHFFFAOYSA-O
SMILES: COC(C)(OC)[NH+](C)C
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)