missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-p-toluidine 98.0+%, TCI America™

Catalog No. p-7126443
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D080725ML 25 mL
D0807500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D080725ML Supplier TCI America Supplier No. D080725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99-97-8
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00008316
InChI Key GYVGXEWAOAAJEU-UHFFFAOYSA-N
Synonym n,n-dimethyl-p-toluidine, dimethyl-p-toluidine, benzenamine, n,n,4-trimethyl, n,n-dimethyl-4-methylaniline, n,n-dimethyl-para-toluidine, 4,n,n-trimethylaniline, dimethyl-4-toluidine, p-dimethylamino toluene, n,n-dimethyl-p-tolylamine, n,n,4-trimethylbenzenamine
PubChem CID 7471
IUPAC Name N,N,4-trimethylaniline
SMILES CN(C)C1=CC=C(C)C=C1

Specifications

Color Yellow
Boiling Point 211°C
Quantity 25 mL
UN Number 2810
Formula Weight 135.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Dimethyl-p-toluidine
EINECSNumber (3)-0191
RTECSNumber XU5803000
TSCA Yes