missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-p-toluidine, 99%

Catalog No. p-120758
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC116905000 500 g Glass bottle
AC116900025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC116905000 Supplier Thermo Scientific Chemicals Supplier No. 116905000
Only null left

CAS: 99-97-8 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-97-8
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00008316
InChI Key GYVGXEWAOAAJEU-UHFFFAOYSA-N
Synonym n,n-dimethyl-p-toluidine, dimethyl-p-toluidine, benzenamine, n,n,4-trimethyl, n,n-dimethyl-4-methylaniline, n,n-dimethyl-para-toluidine, 4,n,n-trimethylaniline, dimethyl-4-toluidine, p-dimethylamino toluene, n,n-dimethyl-p-tolylamine, n,n,4-trimethylbenzenamine
PubChem CID 7471
IUPAC Name N,N,4-trimethylaniline
SMILES CN(C)C1=CC=C(C)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 0.9300g/mL
Boiling Point 211.0°C
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5450 to 1.5470
Linear Formula CH3C6H4N(CH3)2
Quantity 500 g
Beilstein 12, 902
Specific Gravity 0.93
Solubility Information Solubility in water: immiscible
Formula Weight 135.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Dimethyl-p-toluidine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful to aquatic life with long lasting effects.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic in contact with skin.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Use only outdoors or in a well-ventilated area.
Call a
EINECSNumber 202-805-4
RTECSNumber XU5803000
TSCA TSCA
Recommended Storage product darkens on exposure to air, light and moisture

RUO – Research Use Only