Learn More
N,N-Dimethyl-p-toluidine, 99%
CAS: 99-97-8 | C9H13N | 135.21 g/mol
Supplier: Thermo Scientific Chemicals A119880E
Description
N,N-Dimethyl-p-toluidine is used as a polymerization catalyst for polyesters, acrylate and epoxy resins. It is also used as a hardener for dental cements and in adhesives. It serves as an intermediate for photographic chemicals, in industrial glues, in artificial fingernail preparations, colorants, pharmaceuticals. It reacts with vinyl ether in the presence of copper(II) chloride gives tetrahydroquinolines. Further, it is used to accelerate polymerization of ethyl methacrylate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N-Dimethyl-p-toluidine is used as a polymerization catalyst for polyesters, acrylate and epoxy resins. It is also used as a hardener for dental cements and in adhesives. It serves as an intermediate for photographic chemicals, in industrial glues, in artificial fingernail preparations, colorants, pharmaceuticals. It reacts with vinyl ether in the presence of copper(II) chloride gives tetrahydroquinolines. Further, it is used to accelerate polymerization of ethyl methacrylate.
Solubility
Miscible with alcohol, ether and chloroform. Immiscible with water.
Notes
Incompatible with strong oxidizing agents. Store in a cool place.
Specifications
| N,N-Dimethyl-p-toluidine | |
| -25°C | |
| 210°C to 211°C | |
| C9H13N | |
| MFCD00008316 | |
| 1708 | |
| n,n-dimethyl-p-toluidine, dimethyl-p-toluidine, benzenamine, n,n,4-trimethyl, n,n-dimethyl-4-methylaniline, n,n-dimethyl-para-toluidine, 4,n,n-trimethylaniline, dimethyl-4-toluidine, p-dimethylamino toluene, n,n-dimethyl-p-tolylamine, n,n,4-trimethylbenzenamine | |
| GYVGXEWAOAAJEU-UHFFFAOYSA-N | |
| N,N,4-trimethylaniline | |
| 7471 | |
| 99% |
| 99-97-8 | |
| 0.937 | |
| 83°C (181°F) | |
| 1.546 | |
| 2500 g | |
| 774409 | |
| Miscible with alcohol,ether and chloroform. Immiscible with water. | |
| CN(C)C1=CC=C(C)C=C1 | |
| 135.21 | |
| 135.21 |
Chemical Identifiers
| 99-97-8 | |
| 135.21 | |
| GYVGXEWAOAAJEU-UHFFFAOYSA-N | |
| 7471 | |
| CN(C)C1=CC=C(C)C=C1 |
| C9H13N | |
| MFCD00008316 | |
| n,n-dimethyl-p-toluidine, dimethyl-p-toluidine, benzenamine, n,n,4-trimethyl, n,n-dimethyl-4-methylaniline, n,n-dimethyl-para-toluidine, 4,n,n-trimethylaniline, dimethyl-4-toluidine, p-dimethylamino toluene, n,n-dimethyl-p-tolylamine, n,n,4-trimethylbenzenamine | |
| N,N,4-trimethylaniline |
Safety and Handling
GHS H Statement
H301-H311-H331-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
P210-P235-P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P370+P378q-P501c
H227-H301+H311+H331-H373
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 202-805-4
RTECSNumber : XU5803000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only