missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-n-octylamine 97.0+%, TCI America™

Catalog No. p-7126945
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D181425ML 25 mL
D1814500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D181425ML Supplier TCI America Supplier No. D181425ML
Only null left

Chemical Identifiers

CAS 7378-99-6
Molecular Formula C10H23N
Molecular Weight (g/mol) 157.30
MDL Number MFCD00009558
InChI Key UQKAOOAFEFCDGT-UHFFFAOYSA-N
Synonym n,n-dimethyloctylamine, n,n-dimethyl-n-octylamine, dimethyloctylamine, octyldimethylamine, dimethyl octyl amine, n-octyldimethylamine, dimethyl-n-octylamine, 1-octanamine, n,n-dimethyl, octylamine, n,n-dimethyl, unii-20n7h7x4sd
PubChem CID 16224
IUPAC Name dimethyl(octyl)amine
SMILES CCCCCCCCN(C)C

Specifications

Melting Point -57°C
Color Colorless
Boiling Point 90°C
Quantity 25 mL
UN Number 2922
Formula Weight 157.30
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Dimethyl-n-octylamine
EINECSNumber (2)-0176
RTECSNumber RG8075000
TSCA Yes