missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-n-octylamine, 95%

Catalog No. AAA1254630
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1254630 250 g
AAA1254609 10 g
AAA1254618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1254630 Supplier Thermo Scientific Chemicals Supplier No. A1254630
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7378-99-6 | C10H23N | 157.30 g/mol

N,N-Dimethyl-n-octylamine is used as ion-pairing agent in the quantitative estimation of thiazinamium methylsulphate in a pharmaceutical preparation by HPLC. It acts as a counterion in mobile phase for separation of tetracycline, tetracycline analogs and their potential impurities by reversed-phase ion-pair chromatography. Further, it is used in the study of chiral separations by complexation with proteins in capillary zone electrophoresis. In addition to this, it is used as a mobile phase in direct resolution and quantitation of (R)- and (S)-disopyramide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyl-n-octylamine is used as ion-pairing agent in the quantitative estimation of thiazinamium methylsulphate in a pharmaceutical preparation by HPLC. It acts as a counterion in mobile phase for separation of tetracycline, tetracycline analogs and their potential impurities by reversed-phase ion-pair chromatography. Further, it is used in the study of chiral separations by complexation with proteins in capillary zone electrophoresis. In addition to this, it is used as a mobile phase in direct resolution and quantitation of (R)- and (S)-disopyramide

Solubility
Slightly miscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong acids and copper.

Chemical Identifiers

CAS 7378-99-6
Molecular Formula C10H23N
Molecular Weight (g/mol) 157.30
MDL Number MFCD00009558
InChI Key UQKAOOAFEFCDGT-UHFFFAOYSA-N
Synonym n,n-dimethyloctylamine, n,n-dimethyl-n-octylamine, dimethyloctylamine, octyldimethylamine, dimethyl octyl amine, n-octyldimethylamine, dimethyl-n-octylamine, 1-octanamine, n,n-dimethyl, octylamine, n,n-dimethyl, unii-20n7h7x4sd
PubChem CID 16224
IUPAC Name N,N-dimethyloctan-1-amine
SMILES CCCCCCCCN(C)C

Specifications

Melting Point -57°C
Density 0.765
Boiling Point 195°C
Flash Point 65°C (149°F)
Refractive Index 1.4243
Quantity 250 g
UN Number UN2922
Beilstein 1698081
Sensitivity Air Sensitive
Solubility Information Slightly miscible with water.
Formula Weight 157.3
Percent Purity 95%
Chemical Name or Material N,N-Dimethyl-n-octylamine
Show More Show Less
Hazard Category H227-H301-H314
Hazard Statement GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 230-939-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only