missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-m-toluidine 97.0+%, TCI America™

Catalog No. p-7126442
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D080625ML 25 mL
D0806500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D080625ML Supplier TCI America Supplier No. D080625ML
Only null left

Chemical Identifiers

CAS 121-72-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00008305
InChI Key CWOMTHDOJCARBY-UHFFFAOYSA-N
Synonym n,n-dimethyl-m-toluidine, benzenamine, n,n,3-trimethyl, dimethyl-m-toluidine, m-methyl-n,n-dimethylaniline, n,n-dimethyl-m-methylaniline, m,n,n-trimethylaniline, n,n-dimethyl-3-methylaniline, dimetil-m-toluidina, n,n,3-trimethylbenzenamine, benzene, 1-dimethylamino-3-methyl
PubChem CID 8488
IUPAC Name N,N,3-trimethylaniline
SMILES CN(C)C1=CC=CC(C)=C1

Specifications

Color Yellow
Boiling Point 91°C
Quantity 25 mL
UN Number 2810
Formula Weight 135.21
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Dimethyl-m-toluidine
EINECSNumber (3)-0191
RTECSNumber XU5798000
TSCA Yes