missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethyl-3-aminopyrrolidine 96.0+%, TCI America™

Catalog No. D196510G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D196510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D196510G Supplier TCI America Supplier No. D196510G
Only null left

Chemical Identifiers

CAS 64021-83-6
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00059061
InChI Key CZGIEJXGCLWRPY-UHFFFAOYSA-N
PubChem CID 2758520
IUPAC Name N,1-dimethylpyrrolidin-3-amine
SMILES CNC1CCN(C1)C

Specifications

Color Yellow
Boiling Point 167°C
Quantity 10 g
UN Number 2733
Formula Weight 114.19
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N′-Dimethyl-3-aminopyrrolidine
TSCA No
Recommended Storage Refrigerator