accessibility menu, dialog, popup

UserName

CAS RN 64021-83-6

CH3HNNH3C

CAS RN 64021-83-6

IUPAC Name: N,1-dimethylpyrrolidin-3-amine
Synonyms: N,1-dimethylpyrrolidin-3-amine
Molecular Weight (g/mol): 114.19
Molecular Formula: C6H14N2
InChi Key: CZGIEJXGCLWRPY-UHFFFAOYNA-N
SMILES: CNC1CCN(C)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results