missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-2-naphthylamine 98.0+%, TCI America™

Catalog No. D07561G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D07561G 1 g
D07565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D07561G Supplier TCI America Supplier No. D07561G

Chemical Identifiers

CAS 2436-85-3
Molecular Formula C12H13N
Molecular Weight (g/mol) 171.24
MDL Number MFCD00059550
InChI Key IKZPRXHVTFNIEK-UHFFFAOYSA-N
Synonym 2-Dimethylaminonaphthalene
PubChem CID 75527
IUPAC Name N,N-dimethylnaphthalen-2-amine
SMILES CN(C)C1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 46°C
Boiling Point 305°C
Quantity 1 g
Formula Weight 171.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dimethyl-2-naphthylamine