accessibility menu, dialog, popup

UserName

CH3NH3C

CAS RN 2436-85-3

IUPAC Name: N,N-dimethylnaphthalen-2-amine
Synonyms: 2-(dimethylamino)naphthalene
Molecular Weight (g/mol): 171.24
Molecular Formula: C12H13N
InChi Key: IKZPRXHVTFNIEK-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C2C=CC=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results