missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethyl-1-naphthylamine 99.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 274°C |
| Quantity | 500 g |
| Formula Weight | 171.24 |
| Percent Purity | ≥99.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Liquid |
| Chemical Name or Material | N,N-Dimethyl-1-naphthylamine |
Safety and Handling
| RTECSNumber | QM2825000 |
| TSCA | Yes |