missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-1-naphthylamine, 99%

Catalog No. AA36438AC
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36438AC 25 mL
AA36438AA 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA36438AC Supplier Thermo Scientific Chemicals Supplier No. 036438AC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 86-56-6 | C12H13N | 171.243 g/mol

N,N-Dimethyl-1-naphthylamine is used in the nitrate reductase test to form a red precipitate of Prontosil by reacting with a nitrite-sulfanilic acid complex. And is also used to detection and determination of nitrites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86-56-6
Molecular Formula C12H13N
Molecular Weight (g/mol) 171.243
MDL Number MFCD00003919
InChI Key AJUXDFHPVZQOGF-UHFFFAOYSA-N
Synonym n,n-dimethyl-1-naphthylamine, 1-dimethylaminonaphthalene, 1-naphthalenamine, n,n-dimethyl, n,n-dimethyl-1-naphthalenamine, n,n-dimethyl-1-napthylamine, dimethyl 1-naphthyl amine, alpha-dimethylaminonaphthalene, 1-naphthylamine, n,n-dimethyl, dimethyl-alpha-naphthylamine, n,n-dimethyl-1-naftylamin
PubChem CID 6848
IUPAC Name N,N-dimethylnaphthalen-1-amine
SMILES CN(C)C1=CC=CC2=CC=CC=C21

Specifications

Color Yellow
Density 1.039 g/mL
Boiling Point 145°C to 147°C (12 mmHg)
Refractive Index 1.623
Quantity 25 mL
UN Number UN3082
Merck Index 14,3252
Solubility Information Insoluble in water.
Formula Weight 171.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N,N-Dimethyl-1-naphthylamine
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 201-682-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only