missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-1,4-cyclohexanediamine (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. D42881G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42881G 1 g
D42885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42881G Supplier TCI America Supplier No. D42881G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 42389-50-4
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.25
MDL Number MFCD06658434,MFCD20617654
InChI Key GNGZZWTXFNFCMU-UHFFFAOYSA-N
Synonym 1-Amino-4-(dimethylamino)cyclohexane
PubChem CID 345823
IUPAC Name N1,N1-dimethylcyclohexane-1,4-diamine
SMILES CN(C)C1CCC(N)CC1

Specifications

Color Colorless
Boiling Point 205°C
Quantity 1 g
UN Number 2735
Formula Weight 142.25
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Dimethyl-1,4-cyclohexanediamine (cis- and trans- mixture)