missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Dimethyl-1,3-propanediamine 98.0+%, TCI America™
Supplier: TCI America D381125G
Specifications
| N,N′-Dimethyl-1,3-propanediamine | |
| Yellow | |
| C5H16N2 | |
| 25 g | |
| n,n'-dimethyl-1,3-propanediamine, n1,n3-dimethylpropane-1,3-diamine, 1,3-propanediamine, n,n'-dimethyl, 1,3-bis methylamino propane, methyl 3-methylamino propyl amine, 1,3-propanediamine, n1,n3-dimethyl, n,n'-dimethyltrimethylenediamine, n1,n3-dimethyl-1,3-propanediamine, n-3-methylaminopropyl-n-methylamine, n,n'-dimethylpropan-1,3-diamin | |
| C[NH2+]CCC[NH2+]C | |
| 104.20 | |
| 102.18 | |
| Liquid |
| 111-33-1 | |
| 140°C | |
| MFCD00008292 | |
| 2733 | |
| UQUPIHHYKUEXQD-UHFFFAOYSA-P | |
| methyl[3-(methylazaniumyl)propyl]azanium | |
| 66978 | |
| ≥98.0% (T) |
Chemical Identifiers
| 111-33-1 | |
| 104.20 | |
| UQUPIHHYKUEXQD-UHFFFAOYSA-P | |
| 66978 | |
| C[NH2+]CCC[NH2+]C |
| C5H16N2 | |
| MFCD00008292 | |
| n,n'-dimethyl-1,3-propanediamine, n1,n3-dimethylpropane-1,3-diamine, 1,3-propanediamine, n,n'-dimethyl, 1,3-bis methylamino propane, methyl 3-methylamino propyl amine, 1,3-propanediamine, n1,n3-dimethyl, n,n'-dimethyltrimethylenediamine, n1,n3-dimethyl-1,3-propanediamine, n-3-methylaminopropyl-n-methylamine, n,n'-dimethylpropan-1,3-diamin | |
| methyl[3-(methylazaniumyl)propyl]azanium |
Safety and Handling
TSCA : Yes