accessibility menu, dialog, popup

UserName

CAS RN 111-33-1

IUPAC Name: methyl[3-(methylazaniumyl)propyl]azanium
Synonyms: methyl[3-(methylammonio)propyl]azanium
Molecular Weight (g/mol): 104.2
Molecular Formula: C5H16N2
InChi Key: UQUPIHHYKUEXQD-UHFFFAOYSA-P
SMILES: C[NH2+]CCC[NH2+]C
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (3)
  • (3)
  • (4)