missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethyl-1,3-propanediamine 98.0+%, TCI America™

Catalog No. D38115G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38115G 5 g
D381125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38115G Supplier TCI America Supplier No. D38115G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-33-1
Molecular Formula C5H16N2
Molecular Weight (g/mol) 104.20
MDL Number MFCD00008292
InChI Key UQUPIHHYKUEXQD-UHFFFAOYSA-P
Synonym n,n'-dimethyl-1,3-propanediamine, n1,n3-dimethylpropane-1,3-diamine, 1,3-propanediamine, n,n'-dimethyl, 1,3-bis methylamino propane, methyl 3-methylamino propyl amine, 1,3-propanediamine, n1,n3-dimethyl, n,n'-dimethyltrimethylenediamine, n1,n3-dimethyl-1,3-propanediamine, n-3-methylaminopropyl-n-methylamine, n,n'-dimethylpropan-1,3-diamin
PubChem CID 66978
IUPAC Name methyl[3-(methylazaniumyl)propyl]azanium
SMILES C[NH2+]CCC[NH2+]C

Specifications

Color Yellow
Boiling Point 140°C
Quantity 5 g
UN Number 2733
Formula Weight 102.18
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material N,N′-Dimethyl-1,3-propanediamine