missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-1,3-propanediamine 99.0+%, TCI America™

Catalog No. D0790500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0790500ML 500 mL
D079025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0790500ML Supplier TCI America Supplier No. D0790500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-55-7
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.181
MDL Number MFCD00008216
InChI Key IUNMPGNGSSIWFP-UHFFFAOYSA-N
Synonym 3-dimethylaminopropylamine, 3-dimethylamino propylamine, 3-dimethylamino-1-propylamine, n,n-dimethyl-1,3-propanediamine, 1,3-propanediamine, n,n-dimethyl, n,n-dimethyltrimethylenediamine, 3-aminopropyldimethylamine, n,n-dimethylpropane-1,3-diamine, 3-aminopropyl dimethylamine, dimethylaminopropylamine
PubChem CID 7993
IUPAC Name N',N'-dimethylpropane-1,3-diamine
SMILES CN(C)CCCN

Specifications

Color Yellow
Boiling Point 136°C
Quantity 500 mL
UN Number 2734
Formula Weight 102.18
Percent Purity ≥99.0% (GC,T)
Grade Not Available
Physical Form Liquid
Chemical Name or Material N,N-Dimethyl-1,3-propanediamine
EINECSNumber (2)-0158
RTECSNumber TX7525000
TSCA Yes