accessibility menu, dialog, popup

UserName

H3CNCH3NH2

CAS RN 109-55-7

IUPAC Name: (3-aminopropyl)dimethylamine
Synonyms: dimethylaminopropylamine
Molecular Weight (g/mol): 102.18
Molecular Formula: C5H14N2
InChi Key: IUNMPGNGSSIWFP-UHFFFAOYSA-N
SMILES: CN(C)CCCN
Boiling Point 
  • (2)
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)