missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethyl-1,3-propanediamine, 97%

Catalog No. AAA1690106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1690106 5 g
AAA1690114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1690106 Supplier Thermo Scientific Chemicals Supplier No. A1690106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 111-33-1 | C5H16N2 | 104.20 g/mol

Employed as a pharmaceutical intermediate. It is an important raw material used in organic synthesis, agrochemicals and dye stuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a pharmaceutical intermediate. It is an important raw material used in organic synthesis, agrochemicals and dye stuffs.

Solubility
Fully miscible in water, 1e+006 mg/L @ 25°C (est).

Notes
Air sensitive. Store away from oxidizing agents, carbon dioxide and air. Store under inert gas.

Chemical Identifiers

CAS 111-33-1
Molecular Formula C5H16N2
Molecular Weight (g/mol) 104.20
MDL Number MFCD00008292
InChI Key UQUPIHHYKUEXQD-UHFFFAOYSA-P
Synonym n,n'-dimethyl-1,3-propanediamine, n1,n3-dimethylpropane-1,3-diamine, 1,3-propanediamine, n,n'-dimethyl, 1,3-bis methylamino propane, methyl 3-methylamino propyl amine, 1,3-propanediamine, n1,n3-dimethyl, n,n'-dimethyltrimethylenediamine, n1,n3-dimethyl-1,3-propanediamine, n-3-methylaminopropyl-n-methylamine, n,n'-dimethylpropan-1,3-diamin
PubChem CID 66978
IUPAC Name N,N'-dimethylpropane-1,3-diamine
SMILES C[NH2+]CCC[NH2+]C

Specifications

Density 0.817
Boiling Point 143°C to 146°C
Flash Point 32°C (90°F)
Odor Amine-like
Quantity 5 g
UN Number UN2733
Sensitivity Air sensitive; Hygroscopic
Solubility Information Fully miscible in water,1e+006mg/L @ 25°C (est).
Formula Weight 102.18
Percent Purity 97%
Chemical Name or Material N,N'-Dimethyl-1,3-propanediamine
Show More Show Less
Hazard Category H225-H314
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 203-859-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only