missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diisopropylethylenediamine, 97%

CAS: 4013-94-9 | C8H20N2 | 144.26 g/mol

$414.85 - $1279.85

Chemical Identifiers

CAS 4013-94-9
Molecular Formula C8H20N2
Molecular Weight (g/mol) 144.26
MDL Number MFCD00008864
InChI Key MFIGJRRHGZYPDD-UHFFFAOYSA-N
Synonym n,n'-diisopropylethylenediamine, 1,2-ethanediamine, n,n'-bis 1-methylethyl, 1,2-ethanediamine, n1,n2-bis 1-methylethyl, n,n'-diisopropylethylene diamine, n,n'-di propan-2-yl ethane-1,2-diamine, 1,2-bis isopropylamino ethane, n,n'-disopropylethylene diamine, 1,2-ethanediamine,n1,n2-dipropyl, diped, n,n'-diisopropyl-ethylenediamine
PubChem CID 77628
IUPAC Name N,N'-di(propan-2-yl)ethane-1,2-diamine
SMILES CC(C)NCCNC(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1711414
View Documents
Thermo Scientific Chemicals
A1711414
25 g
Each for $414.85
Only null left
 
AAA1711422
View Documents
Thermo Scientific Chemicals
A1711422
100 g
Each for $1,279.85
Only null left
 
Description

Description

N,N'-Diethylethylenediamine is used in the preparation of 2-imidazolidinecarboxaldehydes and 1,4,6,9-tetraalkyl-1,4,6,9-tetraaza-5,10-dioxaperhydroanthracenes by reacting with glyoxal. It acts as a ligand and form coordination complexes with copper(II), which shows thermochromic properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Diethylethylenediamine is used in the preparation of 2-imidazolidinecarboxaldehydes and 1,4,6,9-tetraalkyl-1,4,6,9-tetraaza-5,10-dioxaperhydroanthracenes by reacting with glyoxal. It acts as a ligand and form coordination complexes with copper(II), which shows thermochromic properties.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
Specifications

Chemical Identifiers

4013-94-9
144.26
MFIGJRRHGZYPDD-UHFFFAOYSA-N
77628
CC(C)NCCNC(C)C
C8H20N2
MFCD00008864
n,n'-diisopropylethylenediamine, 1,2-ethanediamine, n,n'-bis 1-methylethyl, 1,2-ethanediamine, n1,n2-bis 1-methylethyl, n,n'-diisopropylethylene diamine, n,n'-di propan-2-yl ethane-1,2-diamine, 1,2-bis isopropylamino ethane, n,n'-disopropylethylene diamine, 1,2-ethanediamine,n1,n2-dipropyl, diped, n,n'-diisopropyl-ethylenediamine
N,N'-di(propan-2-yl)ethane-1,2-diamine

Specifications

4013-94-9
169°C to 171°C
C8H20N2
MFCD00008864
UN2733
Hygroscopic
Miscible with water.
CC(C)NCCNC(C)C
144.26
144.26
N,N'-Diisopropylethylenediamine
0.798
51°C (123°F)
1.4289
25 g
1735571
n,n'-diisopropylethylenediamine, 1,2-ethanediamine, n,n'-bis 1-methylethyl, 1,2-ethanediamine, n1,n2-bis 1-methylethyl, n,n'-diisopropylethylene diamine, n,n'-di propan-2-yl ethane-1,2-diamine, 1,2-bis isopropylamino ethane, n,n'-disopropylethylene diamine, 1,2-ethanediamine,n1,n2-dipropyl, diped, n,n'-diisopropyl-ethylenediamine
MFIGJRRHGZYPDD-UHFFFAOYSA-N
N,N'-di(propan-2-yl)ethane-1,2-diamine
77628
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 223-666-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only