Learn More
N,N'-Diisopropylethylenediamine, 97%
CAS: 4013-94-9 | C8H20N2 | 144.26 g/mol
Supplier: Thermo Scientific Chemicals A1711422
Description
N,N'-Diethylethylenediamine is used in the preparation of 2-imidazolidinecarboxaldehydes and 1,4,6,9-tetraalkyl-1,4,6,9-tetraaza-5,10-dioxaperhydroanthracenes by reacting with glyoxal. It acts as a ligand and form coordination complexes with copper(II), which shows thermochromic properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N′-Diethylethylenediamine is used in the preparation of 2-imidazolidinecarboxaldehydes and 1,4,6,9-tetraalkyl-1,4,6,9-tetraaza-5,10-dioxaperhydroanthracenes by reacting with glyoxal. It acts as a ligand and form coordination complexes with copper(II), which shows thermochromic properties.
Solubility
Miscible with water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
Specifications
| N,N'-Diisopropylethylenediamine | |
| 0.798 | |
| 51°C (123°F) | |
| 1.4289 | |
| 100 g | |
| 1735571 | |
| n,n'-diisopropylethylenediamine, 1,2-ethanediamine, n,n'-bis 1-methylethyl, 1,2-ethanediamine, n1,n2-bis 1-methylethyl, n,n'-diisopropylethylene diamine, n,n'-di propan-2-yl ethane-1,2-diamine, 1,2-bis isopropylamino ethane, n,n'-disopropylethylene diamine, 1,2-ethanediamine,n1,n2-dipropyl, diped, n,n'-diisopropyl-ethylenediamine | |
| MFIGJRRHGZYPDD-UHFFFAOYSA-N | |
| N,N'-di(propan-2-yl)ethane-1,2-diamine | |
| 77628 | |
| 97% |
| 4013-94-9 | |
| 169°C to 171°C | |
| C8H20N2 | |
| MFCD00008864 | |
| UN2733 | |
| Hygroscopic | |
| Miscible with water. | |
| CC(C)NCCNC(C)C | |
| 144.26 | |
| 144.26 |
Chemical Identifiers
| 4013-94-9 | |
| 144.26 | |
| MFIGJRRHGZYPDD-UHFFFAOYSA-N | |
| 77628 | |
| CC(C)NCCNC(C)C |
| C8H20N2 | |
| MFCD00008864 | |
| n,n'-diisopropylethylenediamine, 1,2-ethanediamine, n,n'-bis 1-methylethyl, 1,2-ethanediamine, n1,n2-bis 1-methylethyl, n,n'-diisopropylethylene diamine, n,n'-di propan-2-yl ethane-1,2-diamine, 1,2-bis isopropylamino ethane, n,n'-disopropylethylene diamine, 1,2-ethanediamine,n1,n2-dipropyl, diped, n,n'-diisopropyl-ethylenediamine | |
| N,N'-di(propan-2-yl)ethane-1,2-diamine |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 223-666-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only