missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Diisopropylcarbodiimide 98.0+%, TCI America™
Supplier: TCI America D0254250G
Specifications
| N,N′-Diisopropylcarbodiimide | |
| Yellow | |
| C7H14N2 | |
| 250 g | |
| n,n'-diisopropylcarbodiimide, diisopropylcarbodiimide, 1,3-diisopropylcarbodiimide, n,n-diisopropylcarbodiimide, carbodiimide, diisopropyl, dipcdi, dic, 2-propanamine, n,n'-methanetetraylbis, diisopropylmethanediimine, n,n'-methanediylidenebis propan-2-amine | |
| CC(C)N=C=NC(C)C | |
| 126.203 | |
| CHEBI:53092 | |
| ≥98.0% (GC) |
| 693-13-0 | |
| 147°C | |
| MFCD00065689 | |
| 2929 | |
| BDNKZNFMNDZQMI-UHFFFAOYSA-N | |
| N,N'-di(propan-2-yl)methanediimine | |
| 12734 | |
| 126.20 | |
| Liquid |
Chemical Identifiers
| 693-13-0 | |
| 126.203 | |
| BDNKZNFMNDZQMI-UHFFFAOYSA-N | |
| 12734 | |
| N,N'-di(propan-2-yl)methanediimine |
| C7H14N2 | |
| MFCD00065689 | |
| n,n'-diisopropylcarbodiimide, diisopropylcarbodiimide, 1,3-diisopropylcarbodiimide, n,n-diisopropylcarbodiimide, carbodiimide, diisopropyl, dipcdi, dic, 2-propanamine, n,n'-methanetetraylbis, diisopropylmethanediimine, n,n'-methanediylidenebis propan-2-amine | |
| CHEBI:53092 | |
| CC(C)N=C=NC(C)C |
Safety and Handling
RTECSNumber : FF2175000
TSCA : Yes