missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diisopropylcarbodiimide 98.0+%, TCI America™

Catalog No. p-7126106
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D025425G 25 g
D0254250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D025425G Supplier TCI America Supplier No. D025425G
Only null left

Chemical Identifiers

CAS 693-13-0
Molecular Formula C7H14N2
Molecular Weight (g/mol) 126.203
MDL Number MFCD00065689
InChI Key BDNKZNFMNDZQMI-UHFFFAOYSA-N
Synonym n,n'-diisopropylcarbodiimide, diisopropylcarbodiimide, 1,3-diisopropylcarbodiimide, n,n-diisopropylcarbodiimide, carbodiimide, diisopropyl, dipcdi, dic, 2-propanamine, n,n'-methanetetraylbis, diisopropylmethanediimine, n,n'-methanediylidenebis propan-2-amine
PubChem CID 12734
ChEBI CHEBI:53092
IUPAC Name N,N'-di(propan-2-yl)methanediimine
SMILES CC(C)N=C=NC(C)C

Specifications

Color Yellow
Boiling Point 147°C
Quantity 25 g
UN Number 2929
Formula Weight 126.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N′-Diisopropylcarbodiimide
RTECSNumber FF2175000
TSCA Yes