missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diisopropylaniline, 97%

Catalog No. p-7052716
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5547103 1 g
AAH5547106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5547103 Supplier Thermo Scientific Chemicals Supplier No. H5547103

CAS: 4107-98-6 | C12H19N | 177.291 g/mol

N,N-Diisopropylaniline was used for the synthesis of 4-diisopropylamino benzonitrile. It may be used for the synthesis of 6-(4-bromophenyl)-3-methoxy-5-methyl-8-oxabicyclo[3.2.1]octa-3,6-dien-2-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diisopropylaniline was used for the synthesis of 4-diisopropylamino benzonitrile. It may be used for the synthesis of 6-(4-bromophenyl)-3-methoxy-5-methyl-8-oxabicyclo[3.2.1]octa-3,6-dien-2-one.

Notes
Light Sensitive. Avoid oxidizing agents.

Chemical Identifiers

CAS 4107-98-6
Molecular Formula C12H19N
Molecular Weight (g/mol) 177.291
MDL Number MFCD00048277
InChI Key OVSARSKQWCLSJT-UHFFFAOYSA-N
Synonym n,n-diisopropylaniline, benzenamine, n,n-bis 1-methylethyl, unii-kep9pka41k, kep9pka41k, n,n-dsopropylanlne, diisopropylphenyl amine, acmc-1asew, dsstox_cid_22189, dsstox_rid_79949, dsstox_gsid_42189
PubChem CID 61329
IUPAC Name N,N-di(propan-2-yl)aniline
SMILES CC(C)N(C1=CC=CC=C1)C(C)C

Specifications

Color Colorless to Yellow
Density 0.91 g/mL
Boiling Point 95°C to 96°C (11 mmHg)
Flash Point 84°C (183°F)
Refractive Index 1.519
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 177.29
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N,N-Diisopropylaniline
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only