CAS RN 4107-98-6
IUPAC Name:
N,N-bis(propan-2-yl)aniline
Synonyms:
N,N-diisopropylaniline
Molecular Weight (g/mol):
177.29
Molecular Formula:
C12H19N
InChi Key:
OVSARSKQWCLSJT-UHFFFAOYSA-N
SMILES:
CC(C)N(C(C)C)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
N,N-Diisopropylaniline, 97%
N,N-Diisopropylaniline, 97%
CAS: 4107-98-6 Molecular Formula: C12H19N Molecular Weight (g/mol): 177.291 MDL Number: MFCD00048277 InChI Key: OVSARSKQWCLSJT-UHFFFAOYSA-N Synonym: n,n-diisopropylaniline,benzenamine, n,n-bis 1-methylethyl,unii-kep9pka41k,kep9pka41k,n,n-dsopropylanlne,diisopropylphenyl amine,acmc-1asew,dsstox_cid_22189,dsstox_rid_79949,dsstox_gsid_42189 PubChem CID: 61329 IUPAC Name: N,N-di(propan-2-yl)aniline SMILES: CC(C)N(C1=CC=CC=C1)C(C)C